2-(2,4-Difluorophenyl)-2-methyl-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

17211-1G 17211-25G 17211-5G
76883-644EA 62.6 USD
76883-644 76883-646 76883-648
2-(2,4-Difluorophenyl)-2-methyl-1,3-dioxolane ≥97%
2-(2,4-Difluorophenyl)-2-methyl-1,3-dioxolane
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28129827
CAS Number: 882406-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR