1,4-Difluoro-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

17202-25G 17202-5G
76883-634EA 519.85 USD
76883-634 76883-636
1,4-Difluoro-2-(methoxymethoxy)benzene ≥97%
1,4-Difluoro-2-(methoxymethoxy)benzene
Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09864679
CAS Number: 749230-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR