3-Chloro-5-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Chloro-3-fluoro-5-(trifluoromethyl)benzene

17190-25G 17190-5G
76883-626EA 630.18 USD
76883-626 76883-628
3-Chloro-5-fluorobenzotrifluoride ≥97%
3-Chloro-5-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226526
CAS Number: 1005764-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR