2-(2-Bromo-6-fluoro-3-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

17080-1G 17080-25G 17080-500MG 17080-5G
76883-588EA 337.37 USD
76883-588 76883-590 76883-592 76883-594
2-(2-Bromo-6-fluoro-3-methoxyphenyl)-1,3-dioxolane ≥97%
2-(2-Bromo-6-fluoro-3-methoxyphenyl)-1,3-dioxolane
Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976903
CAS Number: 2221811-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR