2-(2,6-Difluorophenyl)1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

17073-25G 17073-5G
76883-580EA 310.85 USD
76883-580 76883-582
2-(2,6-Difluorophenyl)1,3-dioxolane ≥97%
2-(2,6-Difluorophenyl)1,3-dioxolane
Formula: C₉H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06209063
CAS Number: 467442-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR