1-Bromo-4-iodo-2-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

17062-1G 17062-25G 17062-500MG 17062-5G
76883-564EA 203.7 USD
76883-564 76883-566 76883-568 76883-570
1-Bromo-4-iodo-2-(methylthio)benzene ≥95%
1-Bromo-4-iodo-2-(methylthio)benzene
Formula: C₇H₆BrIS
MW: 328.996 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617269
CAS Number: 1032231-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR