1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-Isobutyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Isobutylpyrazole-4-boronic acid, pinacol ester
17055-25G
17055-5G
76883-556EA
339.49
USD
76883-556
76883-558
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester ≥97%
1-Isobutyl-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₂₃BN₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD05663857 CAS Number: 827614-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...