2,3-Dimethoxy-6-fluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

17039-1G 17039-25G 17039-500MG 17039-5G
76883-542EA 173.99 USD
76883-542 76883-544 76883-546 76883-548
2,3-Dimethoxy-6-fluorophenylboronic acid ≥97% research grade
2,3-Dimethoxy-6-fluorophenylboronic acid
Formula: C₈H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD19105347
CAS Number: 1451392-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR