2,3-Dibromo-5-fluorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2-Dibromo-5-fluoro-3-(trifluoromethyl)benzene

17018-25G 17018-5G
76883-530EA 304.48 USD
76883-530 76883-532
2,3-Dibromo-5-fluorobenzotrifluoride ≥97%
2,3-Dibromo-5-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂Br₂F₄
MW: 321.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800679
CAS Number: 1027511-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR