2-(Benzyloxy)-1,5-dibromo-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

17006-1G 17006-25G 17006-5G
76883-522EA 62.6 USD
76883-522 76883-524 76883-526
2-(Benzyloxy)-1,5-dibromo-3-fluorobenzene ≥97%
2-(Benzyloxy)-1,5-dibromo-3-fluorobenzene
Formula: C₁₃H₉Br₂FO
MW: 360.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976900
CAS Number: 1610471-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR