1,3-Dibromo-2-ethoxy-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

16995-25G 16995-5G
76883-514EA 418 USD
76883-514 76883-516
1,3-Dibromo-2-ethoxy-5-fluorobenzene ≥97%
1,3-Dibromo-2-ethoxy-5-fluorobenzene
Formula: C₈H₇Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD24386128
CAS Number: 1447671-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR