2-(Benzyloxy)-1-bromo-4-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

16766-1G 16766-25G 16766-500MG 16766-5G
76883-474EA 162.32 USD
76883-474 76883-476 76883-478 76883-480
2-(Benzyloxy)-1-bromo-4-(trifluoromethoxy)benzene ≥95%
2-(Benzyloxy)-1-bromo-4-(trifluoromethoxy)benzene
Formula: C₁₄H₁₀BrF₃O₂
MW: 347.127 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28400234
CAS Number: 1704065-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR