2-(Benzyloxy)-3-bromobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16636-1G 16636-25G 16636-500MG 16636-5G
76883-434EA 202.64 USD
76883-434 76883-436 76883-438 76883-440
2-(Benzyloxy)-3-bromobenzaldehyde ≥97%
2-(Benzyloxy)-3-bromobenzaldehyde
Formula: C₁₄H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD09999237
CAS Number: 120980-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR