4-Bromo-2,5-dimethoxytoluene ≥97%

Supplier: AOBChem USA

Warning

16583-25G 16583-5G
76883-426EA 573.95 USD
76883-426 76883-428
4-Bromo-2,5-dimethoxytoluene ≥97%
4-Bromo-2,5-dimethoxytoluene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00666999
CAS Number: 13321-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR