2-(2-Bromo-6-chlorophenyl)-1,3-dioxolane ≥97% research grade

Supplier: AOBChem USA

Warning

16469-1G 16469-25G 16469-500MG 16469-5G
76883-396EA 225.98 USD
76883-396 76883-398 76883-400 76883-402
2-(2-Bromo-6-chlorophenyl)-1,3-dioxolane ≥97% research grade
2-(2-Bromo-6-chlorophenyl)-1,3-dioxolane
Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619377
CAS Number: 2221812-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR