4-Benzyloxy-1-naphthaleneboronic acid 97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Benzyloxynaphthalene-4-boronic acid

16378-1G 16378-25G 16378-5G 16378-500MG
76883-354EA 114.58 USD
76883-354 76883-356 76883-358 77644-742
4-Benzyloxy-1-naphthaleneboronic acid 97%
4-Benzyloxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₅BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD08701761
CAS Number: 183170-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR