2-(2,3-Difluoro-4-iodophenyl)[1,3]dioxolane ≥97%

Supplier: AOBChem USA

Warning

16351-1G 16351-25G 16351-500MG 16351-5G
76883-342EA 225.98 USD
76883-342 76883-344 76883-346 76883-348
2-(2,3-Difluoro-4-iodophenyl)[1,3]dioxolane ≥97%
2-(2,3-Difluoro-4-iodophenyl)[1,3]dioxolane
Formula: C₉H₇F₂IO₂
MW: 312.052 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31619375
CAS Number: 885590-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR