1-Bromo-2-methoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

16022-1G 16022-25G 16022-500MG 16022-5G
76883-298EA 246.13 USD
76883-298 76883-300 76883-302 76883-304
1-Bromo-2-methoxy-3-(trifluoromethyl)benzene ≥97%
1-Bromo-2-methoxy-3-(trifluoromethyl)benzene
Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194485
CAS Number: 871352-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR