2,4-Dichloro-6-fluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
15987-1G
15987-25G
15987-5G
76883-272EA
67.9
USD
76883-272
76883-274
76883-276
2,4-Dichloro-6-fluorobenzoic acid ≥97%
2,4-Dichloro-6-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃Cl₂FO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12404936 CAS Number: 904285-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...