2,4-Dichloro-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15985-1G 15985-25G 15985-500MG 15985-5G
76883-258EA 290.69 USD
76883-258 76883-260 76883-262 76883-264
2,4-Dichloro-6-fluorophenylboronic acid ≥97%
2,4-Dichloro-6-fluorophenylboronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22573056
CAS Number: 1160561-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR