2,3-Dichloro-4-formylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,3-Dichloropyridine-4-carboxaldehyde, 2,3-Dichloroisonicotinaldehyde
15939-1G
15939-25G
15939-5G
76883-250EA
66.84
USD
76883-250
76883-252
76883-254
2,3-Dichloro-4-formylpyridine ≥97%
2,3-Dichloro-4-formylpyridine
Precaución: for Research Use Only
Formula:
C₆H₃Cl₂NO MW: 176.0000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08277282 CAS Number: 884495-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...