2-(4-Fluoro-3-methylphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

15672-1G 15672-25G 15672-5G
76883-164EA 64.72 USD
76883-164 76883-166 76883-168
2-(4-Fluoro-3-methylphenyl)-1,3-dioxolane ≥97%
2-(4-Fluoro-3-methylphenyl)-1,3-dioxolane
Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209312
CAS Number: 773093-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR