4'-Bromo-2',3'-difluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

15660-1G 15660-25G 15660-5G
76883-152EA 109.28 USD
76883-152 76883-154 76883-156
4'-Bromo-2',3'-difluoroacetophenone ≥97%
4'-Bromo-2',3'-difluoroacetophenone
Formula: C₈H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD13192197
CAS Number: 1007346-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR