2-(4-Chloro-2-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

15584-25G 15584-5G
76883-130EA 386.17 USD
76883-130 76883-132
2-(4-Chloro-2-fluorophenyl)-1,3-dioxolane ≥97%
2-(4-Chloro-2-fluorophenyl)-1,3-dioxolane
Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209217
CAS Number: 773102-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR