(E)-5-chloro-2-methoxybenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

155701-1G 155701-25G 155701-500MG 155701-5G
76883-116EA 163.38 USD
76883-116 76883-118 76883-120 76883-122
(E)-5-chloro-2-methoxybenzaldehyde oxime ≥97%
(E)-5-chloro-2-methoxybenzaldehyde oxime
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
Storage Temperature: Ambient
CAS Number: 98545-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR