2-(2,4,6-Trifluorophenyl)-1,3-dioxolane ≥97% research grade

Supplier: AOBChem USA

Warning

15475-25G 15475-5G
76883-086EA 346.92 USD
76883-086 76883-088
2-(2,4,6-Trifluorophenyl)-1,3-dioxolane ≥97% research grade
2-(2,4,6-Trifluorophenyl)-1,3-dioxolane
Formula: C₉H₇F₃O₂
MW: 204.146 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06209156
CAS Number: 773102-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR