2,4,6-Trifluoroanisole ≥97%

Supplier: AOBChem USA

Warning

15467-25G 15467-5G
76883-078EA 222.79 USD
76883-078 76883-080
2,4,6-Trifluoroanisole ≥97%
2,4,6-Trifluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₃O
Storage Temperature: Ambient
MDL Number: MFCD01631502
CAS Number: 219998-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR