1,2-Dibromo-5-chloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

15349-1G 15349-25G 15349-500MG 15349-5G
76883-048EA 296 USD
76883-048 76883-050 76883-052 76883-054
1,2-Dibromo-5-chloro-3-methylbenzene ≥97%
1,2-Dibromo-5-chloro-3-methylbenzene
Formula: C₇H₅Br₂Cl
MW: 284.376 g/mol
Storage Temperature: Ambient
CAS Number: 1000571-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR