1-Fluoro-2-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

15294-25G 15294-5G
76883-040EA 537.88 USD
76883-040 76883-042
1-Fluoro-2-(methoxymethoxy)-4-(trifluoromethyl)benzene ≥95%
1-Fluoro-2-(methoxymethoxy)-4-(trifluoromethyl)benzene
Formula: C₉H₈F₄O₂
MW: 224.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088061
CAS Number: 866615-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR