(E)-4-(2-(dimethylamino)vinyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

152364-1G 152364-25G 152364-500MG 152364-5G
76883-006EA 300.24 USD
76883-006 76883-008 76883-010 76883-012
(E)-4-(2-(dimethylamino)vinyl)benzonitrile ≥97%
(E)-4-(2-(dimethylamino)vinyl)benzonitrile
Formula: C₁₁H₁₂N₂
Storage Temperature: Ambient
CAS Number: 20973-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR