1-Bromo-2,3-dichloro-5-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

15200-1G 15200-25G 15200-5G
76882-986EA 66.84 USD
76882-986 76882-988 76882-990
1-Bromo-2,3-dichloro-5-fluorobenzene ≥95%
1-Bromo-2,3-dichloro-5-fluorobenzene
Formula: C₆H₂BrCl₂F
MW: 243.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878107
CAS Number: 1000577-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR