1,2,3-Tribromobenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
15162-25G
15162-5G
76882-974EA
392.54
USD
76882-974
76882-976
1,2,3-Tribromobenzene ≥97%
1,2,3-Tribromobenzene
Precaución: for Research Use Only
Formula:
C₆H₃Br₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00192668 CAS Number: 608-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...