1-Bromo-2-chloro-4-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

15089-1G 15089-25G 15089-500MG 15089-5G
76882-954EA 211.12 USD
76882-954 76882-956 76882-958 76882-960
1-Bromo-2-chloro-4-methoxy-5-methylbenzene ≥97%
1-Bromo-2-chloro-4-methoxy-5-methylbenzene
Formula: C₈H₈BrClO
MW: 235.506 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28976791
CAS Number: 502159-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR