2,3-Difluoro-4-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
14912-25G
14912-5G
76882-920EA
481.65
USD
76882-920
76882-922
2,3-Difluoro-4-methoxybenzoic acid ≥97%
2,3-Difluoro-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD03428569 CAS Number: 329014-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...