2,4,6-Tribromobenzaldehyde 97%

Supplier: AOBChem USA

Warning

14834-1G 14834-25G 14834-5G 14834-500MG
76882-902EA 79.57 USD
76882-902 76882-904 76882-906 77648-706
2,4,6-Tribromobenzaldehyde 97%
2,4,6-Tribromobenzaldehyde
Formula: C₇H₃Br₃O
Storage Temperature: Refrigerator
MDL Number: MFCD13181607
CAS Number: 45859-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR