(E)-N-(3-fluorophenyl)-2-(hydroxyimino)acetamide ≥97%

Supplier: AOBChem USA

Warning

147853-1G 147853-25G 147853-500MG 147853-5G
76882-886EA 216.43 USD
76882-886 76882-888 76882-890 76882-892
(E)-N-(3-fluorophenyl)-2-(hydroxyimino)acetamide ≥97%
(E)-N-(3-fluorophenyl)-2-(hydroxyimino)acetamide
Formula: C₈H₇FN₂O₂
MW: 182.1517 g/mol
Storage Temperature: Ambient
CAS Number: 1888389-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR