(2-(Benzyloxy)naphthalen-1-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)naphthalen-1-ylboronic acid

14569-1G 14569-25G 14569-500MG 14569-5G
76882-854EA 124.13 USD
76882-854 76882-856 76882-858 76882-860
(2-(Benzyloxy)naphthalen-1-yl)boronic acid ≥95%
(2-(Benzyloxy)naphthalen-1-yl)boronic acid
Formula: C₁₇H₁₅BO₃
MW: 278.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295233
CAS Number: 219834-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR