(E)-1,3-bis(4-bromophenyl)prop-2-en-1-one ≥95%

Supplier: AOBChem USA

Warning

145278-1G 145278-25G 145278-5G 145278-500MG
76882-828EA 79.57 USD
76882-828 76882-830 76882-832 77648-670
(E)-1,3-bis(4-bromophenyl)prop-2-en-1-one ≥95%
(E)-1,3-bis(4-bromophenyl)prop-2-en-1-one

Precaución: for Research Use Only

Formula: C₁₅H₁₀Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00443474
CAS Number: 126443-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR