2-(4-Bromo-2-chlorophenyl)-1,3-dioxolane ≥95%
Supplier: AOBChem USA
|
Warning
|
14257-1G
14257-25G
14257-5G
76882-790EA
71.09
USD
76882-790
76882-792
76882-794
2-(4-Bromo-2-chlorophenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2-chlorophenyl)-1,3-dioxolane
Formula:
C₉H₈BrClO₂ MW: 263.516 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD31619358 CAS Number: 2221812-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...