1-Bromo-3-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

141327-1G 141327-25G 141327-500MG 141327-5G
76882-766EA 217.49 USD
76882-766 76882-768 76882-770 76882-772
1-Bromo-3-((4-methoxybenzyl)oxy)benzene ≥97%
1-Bromo-3-((4-methoxybenzyl)oxy)benzene
Formula: C₁₄H₁₃BrO₂
MW: 293.1558 g/mol
Storage Temperature: Ambient
CAS Number: 944469-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR