2,3-Dimethyl-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

14104-1G 14104-25G 14104-5G 14104-500MG
76882-754EA 102.91 USD
76882-754 76882-756 76882-758 77652-348
2,3-Dimethyl-4-formylphenylboronic acid ≥97%
2,3-Dimethyl-4-formylphenylboronic acid
Formula: C₉H₁₁BO₃
MW: 177.993 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617250
CAS Number: 643093-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR