1-Chloro-2-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

14040-1G 14040-25G 14040-500MG 14040-5G
76882-740EA 201.58 USD
76882-740 76882-742 76882-744 76882-746
1-Chloro-2-(methoxymethoxy)benzene ≥97%
1-Chloro-2-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21101755
CAS Number: 27701-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR