1-Bromo-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

13961-25G 13961-5G
76882-716EA 595.17 USD
76882-716 76882-718
1-Bromo-4-(methoxymethoxy)benzene ≥97%
1-Bromo-4-(methoxymethoxy)benzene
Formula: C₈H₉BrO₂
MW: 217.0599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00457268
CAS Number: 25458-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR