2-Bromo-5-methylanisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2-methoxy-4-methylbenzene

13954-25G 13954-5G
76882-706EA 187.78 USD
76882-706 76882-708
2-Bromo-5-methylanisole ≥95%
2-Bromo-5-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.0605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029185
CAS Number: 95740-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR