1H-Indazole-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

13892-1G 13892-25G 13892-5G
76882-696EA 88.06 USD
76882-696 76882-698 76882-700
1H-Indazole-5-boronic acid ≥97%
1H-Indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06739151
CAS Number: 338454-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR