1-(Methoxymethoxy)-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

13717-25G 13717-5G
76882-670EA 466.8 USD
76882-670 76882-672
1-(Methoxymethoxy)-4-(trifluoromethoxy)benzene ≥97%
1-(Methoxymethoxy)-4-(trifluoromethoxy)benzene
Formula: C₉H₉F₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD30179163
CAS Number: 851341-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR