2-(4-Methoxy-2,3-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,3-Dimethyl-4-methoxyphenylboronic acid pinacol ester

13567-1G 13567-25G 13567-500MG 13567-5G
76882-638EA 203.7 USD
76882-638 76882-640 76882-642 76882-644
2-(4-Methoxy-2,3-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(4-Methoxy-2,3-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₅H₂₃BO₃
Storage Temperature: Ambient
MDL Number: MFCD08706293
CAS Number: 915402-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR