(5-Ethoxy-2-Fluoro-3-Methoxyphenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

134596-1G 134596-25G 134596-500MG 134596-5G
76882-608EA 142.17 USD
76882-608 76882-610 76882-612 76882-614
(5-Ethoxy-2-Fluoro-3-Methoxyphenyl)(Methyl)Sulfane ≥97%
(5-Ethoxy-2-Fluoro-3-Methoxyphenyl)(Methyl)Sulfane
Formula: C₁₀H₁₃FO₂S
MW: 216.2724 g/mol
Storage Temperature: Ambient
CAS Number: 2706961-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR