2-(2-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

13358-1G 13358-25G 13358-500MG 13358-5G
76882-594EA 170.81 USD
76882-594 76882-596 76882-598 76882-600
2-(2-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₆BIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09038529
CAS Number: 857934-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR