1-Bromo-2-Fluoro-5-Methyl-3-(Methylsulfonyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

131791-1G 131791-500MG 131791-5G 131791-25G
76882-582EA 772.34 USD
76882-582 76882-584 76882-586 77657-376
1-Bromo-2-Fluoro-5-Methyl-3-(Methylsulfonyl)Benzene ≥97%
1-Bromo-2-Fluoro-5-Methyl-3-(Methylsulfonyl)Benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₂S
MW: 267.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34829356
CAS Number: 2921865-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR